Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™

Catalog No. AC432250050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432250050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432250050 Supplier Thermo Scientific Chemicals Supplier No. 432250050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1190-16-5
Molecular Formula C5H9Cl2NSi
Molecular Weight (g/mol) 182.12
MDL Number MFCD00019889
InChI Key UIFBMBZYGZSWQE-UHFFFAOYSA-N
Synonym 3-cyanopropyl methyldichlorosilane, 3-cyanopropylmethyldichlorosilane, 4-dichloromethylsilyl-butyronitrile, 3-cyanopropyldichloromethylsilane, butanenitrile, 4-dichloromethylsilyl, unii-a09zd6eimj, 4-dichloromethylsilyl butyronitrile, methylcyanopropyldichlorosilane, 4-dichloro methyl silyl butanenitrile, dichlor-3-kyanpropyl-methylsilan
PubChem CID 14479
IUPAC Name 4-[dichloro(methyl)silyl]butanenitrile
SMILES C[Si](Cl)(Cl)CCCC#N

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 93.0°C to 94.0°C (8.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.15
Formula Weight 182.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyanopropylmethyldichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
EINECSNumber 214-717-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.