Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanopyrazine 99.0+%, TCI America™
SDP

Catalog No. C112325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C112325G 25 g
C1123500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C112325G Supplier TCI America Supplier No. C112325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19847-12-2
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00049361
InChI Key PMSVVUSIPKHUMT-UHFFFAOYSA-N
Synonym pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PubChem CID 73172
ChEBI CHEBI:3982
IUPAC Name pyrazine-2-carbonitrile
SMILES N#CC1=CN=CC=N1

Specifications

Color White
Boiling Point 87°C
Quantity 25 g
Formula Weight 105.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Cyanopyrazine
EINECSNumber (5)-5849
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.