Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanuric chloride, 98%

Catalog No. AAL034420B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL034420B 1000 g
AAL0344218 50 g
AAL0344230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL034420B Supplier Thermo Scientific Chemicals Supplier No. L034420B
Add to Cart

CAS: 108-77-0 | C3Cl3N3 | 184.40 g/mol

Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

Solubility
Soluble in acetone, diethyl ether, dioxane, chloroform, carbon tetrachloride and ketones. Slightly soluble in methanol. Insoluble in cold water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids, alcohols, dimethylformamide, amines and dimethyl sulfoxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
PubChem CID 7954
ChEBI CHEBI:58964
SMILES ClC1=NC(Cl)=NC(Cl)=N1

Specifications

Melting Point 145°C to 149°C
Density 1.92
Boiling Point 192°C to 194°C
Flash Point >190°C (374°F)
Quantity 1000 g
UN Number UN2670
Beilstein 124246
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,diethyl ether,dioxane,chloroform,carbon tetrachloride and ketones. Slightly soluble in methanol. Insoluble in cold water.
IUPAC Name trichloro-1,3,5-triazine
Formula Weight 184.41
Percent Purity 0.98
Chemical Name or Material Cyanuric chloride
Show More Show Less
Hazard Category H302-H314-H317-H330-H335
Hazard Statement GHS H Statement
H330-H314-H302-H317
Fatal if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CYANURIC CHLORIDE
EINECSNumber 203-614-9
RTECSNumber XZ1400000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.