Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanuric chloride, 99%

Catalog No. AC110890051
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110890051 5 kg Plastic drum
AC110892500 250 g Glass bottle
AC110890010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110890051 Supplier Thermo Scientific Chemicals Supplier No. 110890051
Add to Cart

CAS: 108-77-0 | C3Cl3N3 | 184.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
PubChem CID 7954
ChEBI CHEBI:58964
SMILES ClC1=NC(Cl)=NC(Cl)=N1

Specifications

Melting Point 145.0°C to 148.0°C
Color Beige-Brown to Yellow
Density 1.9200g/mL
Boiling Point 190.0°C
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic drum
Quantity 5 kg
Beilstein 26,35
Fieser 03,72; 04,522; 05,687; 06,149; 10,114; 15,105
Specific Gravity 1.92
Merck Index 152691
Solubility Information Solubility in water: reacts. Other solubilities: reacts with ethanol, 250g/L acetone (25°C), soluble in chloroform, hot ether, dioxane, ketones, and carbontetrachloride
IUPAC Name trichloro-1,3,5-triazine
Viscosity 0.57 mPa.s (150°C)
Formula Weight 184.41
Percent Purity 0.99
Physical Form Solidified Mass or Fragments
Chemical Name or Material Cyanuric chloride, 0.99
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
Harmful if swallowed.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-614-9
RTECSNumber XZ1400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.