Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB561 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553228 Supplier Invitrogen™ Supplier No. PA553228
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RADWKRPSQA EEQALSMDFK TLTEGDS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.

Secretory vesicle-specific electron transport protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYB561
Gene Accession No. P49447
Gene Alias CD158F; CD158F1; Cyb561; CYB561A1; cytochrome b561; Cytochrome b-561; cytochrome b561 family, member A1; ferric-chelate reductase 2; FRRS2; KIR2DL5; Mcyt
Gene Symbols CYB561
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYB561. Recombinant protein control fragment (Product #RP-91243).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1534
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.