Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYB5R2 Monoclonal Antibody (OTI4B9)
GREENER_CHOICE

Catalog No. PIMA525631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525631 Supplier Invitrogen™ Supplier No. MA525631
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYB5R2 Monoclonal Antibody for Western Blot, Flow

CYB5R2 is an NADH-cytochrome b5 reductase which are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction. Responsible for NADH-dependent lucigenin chemiluminescence in spermatozoa by reducing both lucigenin and 2-[4-iodophenyl]-3-[4-nitrophenyl]-5-[2,4-disulfophenyl]-2H tetrazolium monosodium salt (WST-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI4B9
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYB5R2
Gene Accession No. Q6BCY4
Gene Alias b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421
Gene Symbols CYB5R2
Host Species Mouse
Immunogen Full length human recombinant protein of CYB5R2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51700
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.