Learn More
Invitrogen™ CYB5R3 Polyclonal Antibody

Description
Immunogen sequence: RPYTPISSDD DKGFVDLVIK VYFKDTHPKF PAGGKMSQYL ESMQIGDTIE FRGPSGLLVY QGKGKFAIRP DKKSNPIIRT VKSVGMIAGG TGITPMLQVI RAIMKDPDDH TVCHLLFANQ TEKDILLRPE LEELRNKHSA RFKLWYTLDR APEAWDYGQG FVNEEMIRDH LPPPEEEPLV LMCGPPPMIQ YACLPNLDHV GHPTERCFVF.
Specifications
Specifications
| Antigen | CYB5R3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.83 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYB5R3 |
| Gene Accession No. | P00387, P20070, Q9DCN2 |
| Gene Alias | 0610016L08Rik; 2500002N19Rik; B5R; C5BR; C85115; CB5R; Cyb5r3; cytochrome b5 reductase 3; DIA1; Dia-1; Diaphorase (NADH) (cytochrome b-5 reductase); diaphorase 1; diaphorase 1 (NADH); Diaphorase1; diaphorase-1; LOC100524254; LOW QUALITY PROTEIN: NADH-cytochrome b5 reductase 3-like; mutant NADH-cytochrome b5 reductase; Nadhcb5; NADH-cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form; NADH-cytochrome b5 reductase 3-like; RNNADHCB5; WU:AL591952.1-001; WU:AL591952.1-002; WU:AL591952.1-003; WU:Cyb5r3 |
| Gene Symbols | CYB5R3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.