Learn More
Fluotics Analytik Jena Cybi Well compatible P60, sterile
Supplier:  Fluotics CYBW60Q.F
Encompass
Analytik Jena Cybi Well compatible P60 filter/sterile Tips in Quadrant only Working Volume 35uL Tip Color Clear Tip Tray 384/60 uL Compare To OL3800-25-935-F 24 racks/case with 96 tips per tray
Catalog No. 50-004-35181