Learn More
Selleck Chemical LLC Tetrahydrozoline HCl S4043-50mg
SDP
Supplier:  Selleck Chemical LLC S404350MG
Encompass_Preferred
Tetrahydrozoline (Tetryzoline) HCl is an imidazoline derivative with alpha receptor agonist activity
Catalog No. 50-310-8693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.