Learn More
Medchemexpress LLC HY-15159 5mg Medchemexpress, Cyclapolin 9 CAS:40533-25-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151595MG
Encompass_Preferred
Medchemexpress, HY-15159 5mg Cyclapolin 9 CAS:40533-25-3 Cyclapolin 9 is a potent, selective and ATP-competitive polo-like kinase 1 (PLK1) inhibitor with an IC50 of 500 nM. Cyclapolin 9 is inactive against other kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.