Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclapolin 9, Tocris Bioscience™

Catalog No. 311610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-161-0 10 mg
31-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-161-0 Supplier R&D Systems Supplier No. 3116/10
This item is not returnable. View return policy

Selective, ATP-competitive PLK1 inhibitor

Selective, ATP-competitive polo-like kinase 1 (PLK1) inhibitor (IC50 = 500 nM).

Chemical Identifiers

CAS 40533-25-3
Molecular Formula C9H4F3N3O4S
Molecular Weight (g/mol) 307.203
InChI Key FHFLXKUKCSDMLD-UHFFFAOYSA-N
Synonym cyclapolin 9, 7-nitro-3-oxy-5-trifluoromethyl-benzothiazole-2-carboxylic acid amide, 7-nitro-5-trifluoromethyl-2-benzothiazolecarboxamide-3-oxide, cyclapolin-9, 2-carbamoyl-7-nitro-5-trifluoromethyl-1,3-benzothiazol-3-ium-3-olate, 7-nitro-5-trifluoromethyl-2-benzothiazolecarboxamide 3-oxide, 7-nitro-3-oxido-5-trifluoromethyl-1,3-benzothiazol-3-ium-2-carboxamide, 2-aminocarbonyl-7-nitro-5-trifluoromethyl-1,3-benzothiazol-3-ium-3-olate
PubChem CID 2779952
IUPAC Name 7-nitro-3-oxido-5-(trifluoromethyl)-1,3-benzothiazol-3-ium-2-carboxamide
SMILES C1=C(C=C(C2=C1[N+](=C(S2)C(=O)N)[O-])[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 307.21
Percent Purity >98%
Chemical Name or Material Cyclapolin 9
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.