Learn More
Enzo Life Sciences cGMP ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901013
Encompass_Preferred
Most sensitive (0.088 pmol/ml) colorimetric kit  for extracellular cGMP quantification. This is a colorimetric competitive immunoassay kit for the quantitative determination of extracellular cGMP. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error. Alternative Name: cyclic GMP. Sensitivity: (non-acetylated) 0.37 pmol/ml; (acetylated) 0.088 pmol/ml (range (non-acetylated) 0.16 - 500 pmol/ml; (acetylated) 0.16 - 100 pmol/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer 2 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine
Catalog No. 50-658-205
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.