Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595251 Supplier Invitrogen™ Supplier No. PA595251
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human Cyclin A2 shares 74.5% amino acid (aa) sequence identity with mouse Cyclin A2. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human HL-60 whole cell, human MCF-7 whole cell, rat stomach tissue, rat testis tissue, mouse stomach tissue. IHC: human colorectal adenocarcinoma tissue. ICC/IF: A431 cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdks) and they control transition at different specific phases of the cell cycle. The temporal expression of cyclins is tightly regulated and subsequently plays a critical role in controlling the enzymatic activity of cdks. These cyclin / cdk complexes are essential for passage through specific stages in the cell cycle. In mammalian somatic cells, cyclin A is required for S phase and passage through G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin A2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CCNA2
Gene Accession No. P20248, P51943
Gene Alias AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg
Gene Symbols CCNA2
Host Species Rabbit
Immunogen E.coli-derived human Cyclin A2 recombinant protein (Position: A10-K168).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114494, 12428, 890
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.