Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537254 Supplier Invitrogen™ Supplier No. PA537254
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 158 kDa. Purity is >95% by SDS-PAGE.

Cell proliferation is controlled at specific stages of the cell cycle by distinct protein kinase complexes. These complexes consist of a catalytic subunit associating with a specific regulatory subunit to form the active kinase. The cyclins, which include cyclin A, B, C, D, E, F, G, H, I, K, L, T and their related proteins, including Dbf4, comprise the regulatory subunits of these kinase complexes. The controlled activation of the kinase complexes at various intervals of the cell cycle is regulated by the availability of the cyclins to the catalytic subunit. Unlike the catalytic subunit, which is expressed continually, the expression and stability of the regulatory subunit fluctuates depending on the stage of the cell cycle, thereby regulating kinase activity. Cyclin B3, also known as CCNB3 or CYCB3, is a 1,395 amino acid nuclear protein that belongs to the cyclin family of regulatory proteins. Expressed in testis with lower expression in a variety of other tissues, cyclin B3 is thought to be required for early meiotic prophase I, playing an important role in the meiotic cell cycle. Three isoforms of cyclin B3 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CCNB3
Gene Accession No. Q810T2, Q8WWL7
Gene Alias CCNB3; CYCB3; cyclin B3; CyclinB3; G2/mitotic-specific cyclin-B3; Protein Ccnb3; RGD1564367
Gene Symbols CCNB3
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide encompassing a sequence within the center region of human Cyclin B3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209091, 317389, 85417
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.