Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody

Catalog No. RM9104S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9104S 1 mL Concentrated
RM9104S0 100 μL Concentrated
RM9104RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RM9104S Supplier Epredia™ Supplier No. RM9104S

missing translation for 'validMainframeMsg'

Obtain accurate and reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody.

Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Rabbit

Clone: SP4

Isotype: IgG

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells, breast CA or mantle cell lymphoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Cyclin D1/Bcl-1
Applications Western Blot
Classification Monoclonal
Clone SP4
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human cyclin D1
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.