Learn More
Invitrogen™ Cyclin D1 Recombinant Rabbit Monoclonal Antibody (SP4)
Shop All Thermo Scientific Products

Description
MA5-14512 targets Cyclin D1/Bcl-1 in IHC (P), ICC/IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-14512 immunogen is a synthetic peptide from C-terminus of human cyclin D1.
Specifications
Specifications
| Antigen | Cyclin D1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | SP4 |
| Concentration | 0.046 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.2-7.4 |
| Gene | CCND1 |
| Gene Accession No. | P24385, P25322, P39948 |
| Gene Alias | AI327039; B-cell CLL/lymphoma 1; B-cell lymphoma 1 protein; BCL1; BCL-1; BCL-1 oncogene; CCND1; CCND1 protein; cD1; cDNA; CycD1; cyclin D1; Cyl-1; D11S287E; FLJ93625; G1/S-specific cyclin-D1; mRNA; PRAD1; PRAD-1; PRAD1 oncogene; U21B31 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.