Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclin E Monoclonal Antibody (HE12)
GREENER_CHOICE

Catalog No. 321600
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-160-0 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-160-0 Supplier Invitrogen™ Supplier No. 321600
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Cyclin E Monoclonal Antibody for Western Blot, ICC/IF, IHC, IP

Cyclin E belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. Cyclin E forms a complex with and functions as a regulatory subunit of CDK2, whose activity is required for cell cycle G1/S transition. This protein accumulates at the G1-S phase boundary and is degraded as cells progress through S phase. Overexpression of this gene has been observed in many tumors, which results in chromosome instability, and thus may contribute to tumorigenesis. This protein was found to associate with, and be involved in, the phosphorylation of NPAT protein (nuclear protein mapped to the ATM locus), which participates in cell-cycle regulated histone gene expression and plays a critical role in promoting cell-cycle progression in the absence of pRB. Two alternatively spliced transcript variants of this gene, which encode distinct isoforms, have been described. Two additional splice variants were reported but detailed nucleotide sequence information is not yet available.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin E
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HE12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CCNE1
Gene Accession No. P24864
Gene Alias AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1
Gene Symbols CCNE1
Host Species Mouse
Immunogen Full-length, recombinant, human cyclin E protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 898
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.