Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin-E Monoclonal Antibody (ZM121), MonoMab™, Zeta

Catalog No. p-7238778
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-6155 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-6155 Supplier ZETA CORPORATION Supplier No. Z2431ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Cocktail

A recommended positive control tissue for this product is ZMTB-1, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

Cyclin-E Monoclonal antibody specifically detects Cyclin-E in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Cyclin-E
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZM121
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene CCNE1
Gene Accession No. P24864
Gene Alias AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1
Host Species Mouse
Immunogen Recombinant human Cyclin E (CCNE1) protein fragment (around aa 10-176)
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 898
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.