Learn More
Invitrogen™ Cyclin H Monoclonal Antibody (33TF2-2D4)

Description
MA3-025 has been used successfully in Western blotting, Immunoprecipitation and Immunofluorescence applications to detect Cyclin H in human samples.
Specifications
Specifications
| Antigen | Cyclin H |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 33TF2-2D4 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.05% sodium azide |
| Gene | CCNH |
| Gene Accession No. | P51946 |
| Gene Alias | 6330408H09Rik; AI661354; AV102684; AW538719; CAK; CAK complex subunit; Ccnh; CDK-activating kinase complex subunit; CycH; cyclin H; cyclin-dependent kinase-activating kinase complex subunit; cyclin-H; MO15-associated protein; OTTHUMP00000222395; OTTHUMP00000222396; p34; p37 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.