Learn More
Bachem Cyclo(-Arg-Gly-Asp-D-Phe-Val)
SDP
Supplier:  Bachem 4026200.0001
Encompass_Preferred
Cyclo(-Arg-Gly-Asp-D-Phe-Val) Trifluoroacetate, 137813-35-5, C26H38N8O7, Mr 574.64, 1mg. c(RGDfV), EMD 66203, c(Arg-Gly-Asp-DPhe-Val). c(RGDfV) is a potent inhibitor of cell adhesion. It inhibited tumor cell adhesion to laminin and vitronectin substrates. In vivo investigations in rats provided the first evidence of its utility in the amelioration of ischemic acute renal failure. c(RGDfV) inhibited tubular obstruction by preventing cell-cell adhesion. Hammes et al. showed that the cyclopeptide can also be used to prevent retinal neovascularization. c(RGDfV) binds to avb3 receptors.
Catalog No. 50-259-626
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.