Learn More
CPC Scientific Cyclo(-Arg-Ala-Asp-D-Phe-Glu-) (ammonium salt) 0.5MG
SDP
Supplier:  CPC Scientific RGDP002A
Encompass
This peptide is a negative control for the cyclo (-RGDfE) peptide. RGD peptides are modulators of cell adhesion, and are recognized by several members of the integrin family. This peptide has a low affinity binding to integrin peptides.SEQUENCE: Cyclo(-Arg-Ala-Asp-D-Phe-Glu-) (ammonium salt)ONE-LETTER SEQUENCE: Cyclo(-RADfE-)MOLECULAR FORMULA: MOLECULAR WEIGHT: 618.65STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [756500-23-9]SYNONYMS: RESEARCH AREA: CancerCATEGORY: Fibronectin and RGD Sequences »REFERENCES:Wester, H. et al. J. Nucl. Med. 46, 1940 (2005)Janssen, M. et al. Canc. Res. 62, 6146 (2002)SKU(s): RGDP-002A, RGDP-002B, RGDP-002C
Catalog No. 50-211-0373
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.