Learn More
Apexbio Technology LLC Cyclo (-RGDfC), 100mg CAS# 862772-11-0
SDP
Supplier:  Apexbio Technology LLC A8790100
Encompass_Preferred
RGD tumor targeting peptide can bind to v3 receptor. c (RGDfC) can be easily conjugated to convistatin or drug surface. Cyclo (-RGDfC) is an integrin avb3-affine polypeptide, a circular RGDfc sequence. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8402
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.