Learn More
TARGETMOL CHEMICALS INC Cyclo-RGDfK 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T68125MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cyclo(-RGDfK) is a potent and selective inhibitor of integrin alphavbeta3 with IC50 of 0.94 nM. Cyclo specifically targets tumor microvasculature and cancer cells by binding to alphavbeta3 integrin on the cell surface. Purity 99.12%
Catalog No. 50-287-7344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.