Learn More
Medchemexpress LLC HY-P0023 100mg , Cyclo(-RGDfK) CAS:161552-03-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYP0023100MG
Encompass_Preferred
Medchemexpress, HY-P0023 100mg Cyclo(-RGDfK) CAS:161552-03-0 Formula:C27H41N9O7 IC50: 0.94 nM (α v β 3 integrin) . Purity:98% Cyclo(-RGDfK) is a potent and selective inhibitor of the α v β 3 integrin, with a IC 50 of 0.94 nM. Sequence: Cyclo(Arg-Gly-Asp-Phe-Lys). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3009
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.