Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclodecane, MP Biomedicals

Catalog No. ICN226743 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226743 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226743 Supplier MP Biomedicals, Inc Supplier No. 0522674301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 293-96-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
InChI Key LMGZGXSXHCMSAA-UHFFFAOYSA-N
Synonym cyclodecan, cyclodecyl, acmc-1cklb, cyclodecane
PubChem CID 9267
IUPAC Name cyclodecane
SMILES C1CCCCCCCCC1

Specifications

Quantity 10 g
Chemical Name or Material Cyclodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.