Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclodecane 94.0+%, TCI America™
SDP

Catalog No. C14701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14701G Supplier TCI America Supplier No. C14701G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 293-96-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00003711
InChI Key LMGZGXSXHCMSAA-UHFFFAOYSA-N
Synonym cyclodecan, cyclodecyl, acmc-1cklb, cyclodecane
PubChem CID 9267
IUPAC Name cyclodecane
SMILES C1CCCCCCCCC1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 201°C
Quantity 1 g
Formula Weight 140.27
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.