Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinyltetramethyldisiloxane, 97%

Catalog No. AC249210500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC249210500 50 g Glass bottle
AC249210100 10 g Glass bottle
AC249212500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC249210500 Supplier Thermo Scientific Chemicals Supplier No. 249210500
Only null left
Add to Cart

CAS: 2627-95-4 | C8H18OSi2 | 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-95-4
Molecular Formula C8H18OSi2
Molecular Weight (g/mol) 186.40
MDL Number MFCD00014933
InChI Key BITPLIXHRASDQB-UHFFFAOYSA-N
Synonym 1,3-divinyltetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, disiloxane, 1,3-diethenyl-1,1,3,3-tetramethyl, 1,1'-divinyltetramethyldisiloxane, unii-sj4d71zqf4, bisvinyltetramethyldisiloxane, 1,3-divinyl-1,1,3,3-tetramethyl disiloxane, sym-tetramethyldivinyldisiloxane, tetramethyl-1,3-divinyldisiloxane, 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane
PubChem CID 75819
IUPAC Name ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Melting Point -99.7°C
Color Colorless
Density 0.8130g/mL
Boiling Point 139°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4110 to 1.413
Linear Formula [H2C=CHSi(CH3)2]2O
Quantity 50 g
Specific Gravity 0.813
Solubility Information Solubility in water: insoluble
Formula Weight 186.4
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Divinyltetramethyldisiloxane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-099-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.