Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Cyclodextrin, MP Biomedicals™

Catalog No. ICN19005301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19005301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19005301 Supplier MP Biomedicals, Inc Supplier No. 0219005301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Density 1.624g/cm3 (L)
Quantity 1 g
Formula Weight 1134.9
Physical Form Solid
Chemical Name or Material β-Cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.