CAS RN 7585-39-9
beta-Cyclodextrin, 98%
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
beta-Cyclodextrin
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
R&D Systems™ b-Cyclodextrin
A Tocris product, part of R&D Systems. Cyclic complexing agent
β-Cyclodextrin, >98%, MP Biomedicals™
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
beta-Cyclodextrin 98.0+%, TCI America™
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
beta-Cyclodextrin 99.0+%, TCI America™
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Sigma Aldrich Beta-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Beta-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Alpha-Cyclodextrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Tris(2-Carboxyethyl)Phosphine Hydrochloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
β-Cyclodextrin, MP Biomedicals™
CAS: 7585-39-9 Molecular Formula: C42H70O35 Molecular Weight (g/mol): 1134.99 MDL Number: MFCD00078139 InChI Key: WHGYBXFWUBPSRW-UHFFFAOYNA-N Synonym: cycloheptaamylose,cycloheptapentylose,.beta.-cyclodextrin,caraway,cyclo-hepta-amylose,cycloheptamaltose,.beta.-dextrin,beta-cycloamylose,beta-cyclodextrin,.beta.-cycloamylose PubChem CID: 131707246 IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O