Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclododecane epoxide, mixture of cis- andtrans-isomers, Thermo Scientific™

Catalog No. p-2691260
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110950250 25 g
AC110951000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110950250 Supplier Thermo Scientific Chemicals Supplier No. 110950250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286-99-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.31
MDL Number MFCD00005116
InChI Key VLJLXEKIAALSJE-UHFFFAOYNA-N
Synonym 1,2-epoxycyclododecane, 13-oxabicyclo 10.1.0 tridecane, epoxycyclododecane, cyclododecene oxide, cyclododecene epoxide, cyclododecane epoxide, cyclododecane, 1,2-epoxy, ccris 3754, cyclododecane oxide
PubChem CID 9248
IUPAC Name 13-oxabicyclo[10.1.0]tridecane
SMILES C1CCCCCC2OC2CCCC1

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4795 to 1.4815 (20°C, 589nm)
Quantity 25 g
Formula Weight 182.31
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Cyclododecane epoxide, mixture of cis- and trans-isomers, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.