Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclododecane 99.0+%, TCI America™
SDP

Catalog No. C0554100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0554100G 100 g
C055425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0554100G Supplier TCI America Supplier No. C0554100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 294-62-2
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00014258
InChI Key DDTBPAQBQHZRDW-UHFFFAOYSA-N
PubChem CID 9268
IUPAC Name cyclododecane
SMILES C1CCCCCCCCCCC1

Specifications

Melting Point 63°C
Color White
Boiling Point 111°C
Quantity 100 g
Formula Weight 168.32
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclododecane
EINECSNumber (3)-2240
RTECSNumber GV2295000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.