Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheptatriene 90%, Thermo Scientific™

Catalog No. AC111050250
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111050250 25 mL
AC111052500 250 mL
AC111050010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111050250 Supplier Thermo Scientific Chemicals Supplier No. 111050250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-25-2
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
InChI Key CHVJITGCYZJHLR-UHFFFAOYSA-N
Synonym cycloheptatriene, tropilidene, 1,3,5-cycloheptatriene, tropilidine, tropiliden, cycloheptatrien, tropyliden, zykloheptatrien, 1h-7 annulene, tropilidin
PubChem CID 11000
ChEBI CHEBI:37519
IUPAC Name cyclohepta-1,3,5-triene
SMILES C1C=CC=CC=C1

Specifications

Color Brown to Yellow
Infrared Spectrum Authentic
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Quantity 25 mL
Beilstein 05, 280
Fieser 01,177
Formula Weight 92.14
Percent Purity ≥88%
Physical Form Liquid
Chemical Name or Material Cycloheptatriene, 90%
missing translation for 'healthHazard1' LACHRYMATOR
EINECSNumber 208-866-3
RTECSNumber GU3675000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.