Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanemethanol 98.0+%, TCI America™
SDP

Catalog No. C0595100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
C0595100G 100 g
C059525G 25 g
C0595500G 500 g
3 options

Catalog No. C0595100G

Supplier: TCI America C0595100G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-49-2
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00001510
InChI Key VSSAZBXXNIABDN-UHFFFAOYSA-N
Synonym cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane
PubChem CID 7507
IUPAC Name cyclohexylmethanol
SMILES C1CCC(CC1)CO

Specifications

Color Colorless
Boiling Point 183°C
Quantity 100 g
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexanemethanol
RTECSNumber GV5075000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.