Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanemethylamine 98.0+%, TCI America™
SDP

Catalog No. A1114100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1114100ML 100 mL
A111425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1114100ML Supplier TCI America Supplier No. A1114100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3218-02-8
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001520
InChI Key AVKNGPAMCBSNSO-UHFFFAOYSA-N
Synonym cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049
PubChem CID 76688
IUPAC Name cyclohexylmethanamine
SMILES C1CCC(CC1)CN

Specifications

Color Yellow
Boiling Point 163°C
Quantity 100 mL
UN Number 2734
Formula Weight 113.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexanemethylamine
RTECSNumber GV6128000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.