Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanol, 97%, Spectrum™ Chemical
SDP

Catalog No. 18610101 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-101 500 mL Steel Can
18-610-100 4 L Steel Can
18-610-098 20 L Steel Can
18-610-099 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-101 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1460500MLGL

Chemical Identifiers

CAS 110-82-7
Molecular Weight (g/mol) 100.16

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Steel Can
Boiling Point 161.84°C
Percent Purity 97%
Grade Reagent
Chemical Name or Material Cyclohexanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.