Learn More
Sigma Organic Chemistry Cyclohexanone | 17L | 108-94-1 | MFCD00001625
Supplier: Sigma Organic Chemistry C10218017L
Cyclohexanone, 17L
About This Item:
Linear Formula: C6H10(=O)
Molecular Weight: 98.14
CAS Number: 108-94-1
MDL Number: MFCD00001625
UNSPSC Code: 12352100
Purity: 0.998
Boiling Point: 155 C (lit.)
Melting Point: -47 C (lit.)
Density: 0.947 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.450 (lit.)
Storage: room temp
EINECS Number: 203-631-1
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.