Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002678 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-678 4 L Narrow Mouth Amber Glass Bottle
02-002-679 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-002-678 Supplier Avantor Supplier No. 921003
Add to Cart
Add to Cart

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Quantity 4 L
Flash Point 44.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.94kg
Formula Weight 98.14
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.