Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexene oxide, 98+%

Catalog No. aaa1318522
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1318522 100 g
AAA1318536 500 g
AAA131850E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1318522 Supplier Thermo Scientific Chemicals Supplier No. A1318522
Only null left
Add to Cart

CAS: 286-20-4 | C6H10O | 98.145 g/mol

It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

Solubility
Insoluble in water

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286-20-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00005162
InChI Key ZWAJLVLEBYIOTI-UHFFFAOYSA-N
Synonym cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide
PubChem CID 9246
IUPAC Name 7-oxabicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)O2

Specifications

Melting Point -40°C
Density 0.972
Boiling Point 129°C to 130°C
Flash Point 24°C (75°F)
Refractive Index 1.452
Quantity 100 g
UN Number UN2920
Beilstein 383568
Solubility Information Insoluble in water
Formula Weight 98.15
Percent Purity ≥98%
Chemical Name or Material Cyclohexene oxide
Show More Show Less
Hazard Category H226-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H311-H314-H318-H226-H302-H332
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 206-007-7
RTECSNumber RN7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.