Learn More
Bioworld Cycloheximide, 5 g
Supplier:  Bioworld 403100282
Encompass_Preferred
Synonym: Naramycin A; Actidione; 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimideAn antibacterial protein synthesis inhibitorAppearance: SolidBoiling Point : 491.79 °C at 760 mmHg (Predicted)Density : 1.14 g/cm3 (Predicted)IUPAC_Name: 4-[2-(3,5-Dimethyl-2-Oxo-Cyclohexyl)-2-Hydroxyethyl]-2,6-PiperidinedioneMelting Point: 110 °CRTECS Number: MA4375000
Catalog No. 50-255-724