Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheximide, from Microbial, MP Biomedicals™

Catalog No. ICN10018305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10018305 5 g
ICN10018301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10018305 Supplier MP Biomedicals, Inc Supplier No. 0210018305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Color White
Assay Percent Range ≥94%
Quantity 5 g
Formula Weight 281.35
Physical Form Powder
Chemical Name or Material Cycloheximide, from Microbial; ∽800μg/mg
EINECSNumber 200-636-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.