CAS RN 66-81-9
Thermo Scientific Chemicals Cycloheximide, 95%
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: cycloheximide,actidione,cycloheximide,naramycin a,kaken,actidion,actidone,hizarocin,naramycin,neocycloheximide PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Thermo Scientific Chemicals Cycloheximide, ultrapure, 95%
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Cycloheximide solution, 100mg/mL in DMSO, 0.2 micron filtered
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: CHX PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Cycloheximide, from Microbial, MP Biomedicals™
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: cycloheximide,actidione,cycloheximide,naramycin a,kaken,actidion,actidone,hizarocin,naramycin,neocycloheximide PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Thermo Scientific Chemicals Cycloheximide, 90%
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Cycloheximide, MP Biomedicals™
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: Acti-Dione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl] glutarimide, Naramycin A, Actidione, CHX PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Cycloheximide from Microbial, MP Biomedicals
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: cycloheximide,actidione,cycloheximide,naramycin a,kaken,actidion,actidone,hizarocin,naramycin,neocycloheximide PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
R&D Systems™ Cycloheximide
CAS: 66-81-9 Molecular Formula: C15H23NO4
Cycloheximide, High Purity, MilliporeSigma™
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
MilliporeSigma™ Cycloheximide, InSolution™, Calbiochem™,
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: cycloheximide,actidione,cycloheximide,naramycin a,kaken,actidion,actidone,hizarocin,naramycin,neocycloheximide PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
MilliporeSigma™ Cycloheximide, ≥90%, Calbiochem™,
CAS: 66-81-9 Molecular Formula: C15H23NO4 Molecular Weight (g/mol): 281.35 MDL Number: MFCD00082346 InChI Key: YPHMISFOHDHNIV-QTEFRXOUNA-N Synonym: cycloheximide,actidione,cycloheximide,naramycin a,kaken,actidion,actidone,hizarocin,naramycin,neocycloheximide PubChem CID: 6197 ChEBI: CHEBI:27641 IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione SMILES: [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1