Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheximide, MP Biomedicals™

Catalog No. MP219452725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219452725 25 g
MP219452701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219452725 Supplier MP Biomedicals, Inc Supplier No. 0219452725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cycloheximide is an antibiotic which is very active against many molds, yeasts, and phytopathogenic fungi.

Cycloheximide exhibits somewhat lower activity against bacteria and certain fungi. Control of various molds and fungi in gelatin-based photographic emulsions, photoengraving glues, and other light-sensitive products is suggested.

  • Cycloheximide (CHX) is an antibiotic produced by S. griseus. Its main biological activity is translation inhibition in eukaryotes resulting in cell growth arrest and cell death.
  • CHX is widely used for selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors.
  • Cycloheximide inhibits peptide synthesis in eukaryotic organisms but not in prokaryotes. Protein synthesis is blocked by the interaction of cycloheximide with the translocase enzyme. This interaction prohibits the translocation of messenger RNA on the cytosolic, 80S ribosomes without inhibiting organelle protein synthesis.
  • Cycloheximide is known to induce FAS/FAS Ligand apoptosis, and triggers apoptosis in HL-60 cells, T-cell hybridomas, Burkitt's lymphoma cells in addition to a variety of other cell types. Cycloheximide will also delay or inhibit apoptosis induced by other agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym Acti-Dione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl] glutarimide, Naramycin A, Actidione, CHX
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Physical Form Powder
Melting Point 103°C to 115°C
Color Off-White
Quantity 25 g
Formula Weight 281.36g/mol
Grade Cell Culture
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.