Learn More
Sigma Aldrich Fine Chemicals Biosciences Cycloheximide solution, 66-81-9, MFCD00082346, 1 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences C48591ML
Empirical Formula (Hill Notation): C15H23NO4, Molecular Weight: 281.35, Synonym: CHX, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered. Tested for cell growth arrest, selection of cycloheximide resistant yeast, apoptosis induction, and facilitation of apoptosis induction by FasL.