Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyl Vinyl Ether (stabilized with KOH) 98.0+%, TCI America™
SDP

Catalog No. C088525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088525ML 25 mL
C0885500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C088525ML Supplier TCI America Supplier No. C088525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2182-55-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00046358
InChI Key VGIYPVFBQRUBDD-UHFFFAOYSA-N
Synonym cyclohexyl vinyl ether, vinyloxy cyclohexane, cyclohexane, ethenyloxy, vinylcyclohexyl ether, cyclohexylvinylether, vinyl cyclohexyl ether, ethenyloxy cyclohexane, vinyloxycyclohexane, cyclohexylvinyl ether, vinyloxy cyclohexane #
PubChem CID 75129
IUPAC Name ethenoxycyclohexane
SMILES C=COC1CCCCC1

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 mL
UN Number 3271
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Vinyl Ether (stabilized with KOH)
EINECSNumber (3)-3687
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.