Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C21885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2188-5G 5 g
C21881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2188-5G Supplier TCI America Supplier No. C21885G

Chemical Identifiers

CAS 4441-56-9
Molecular Formula C6H13BO2
MDL Number MFCD01311824
InChI Key XDRVAZAFNWDVOE-UHFFFAOYSA-N
PubChem CID 199578
IUPAC Name cyclohexylboronic acid

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Synonym cyclohexylboronicacid, cyclohexaneboronic acid, boronic acid, cyclohexyl, cyclohexyl boronic acid, boronic acid, cyclohexyl-9 ci
SMILES OB(O)C1CCCCC1
Molecular Weight (g/mol) 127.98
Formula Weight 127.98
Physical Form Crystalline Powder
RTECSNumber GU7171000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.