Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexylmagnesium Bromide (ca. 18% in Tetrahydrofuran, ca. 1mol/L), TCI America™
SDP

Catalog No. c1504100g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1504100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1504100G Supplier TCI America Supplier No. C1504100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 931-50-0
Molecular Formula C6H11BrMg
Molecular Weight (g/mol) 187.363
MDL Number MFCD01321152
InChI Key GFTXWCQFWLOXAT-UHFFFAOYSA-M
PubChem CID 11805477
IUPAC Name magnesium;cyclohexane;bromide
SMILES C1CC[CH-]CC1.[Mg+2].[Br-]

Specifications

Quantity 100 g
UN Number 2924
Formula Weight 187.36
Chemical Name or Material Cyclohexylmagnesium Bromide (ca. 18% in Tetrahydrofuran, ca. 1mol/L)
EINECSNumber (3)-3901
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.