Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyltrichlorosilane 98.0+%, TCI America™
SDP

Catalog No. C089225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089225G 25 g
C08925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089225G Supplier TCI America Supplier No. C089225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-12-4
Molecular Formula C6H11Cl3Si
Molecular Weight (g/mol) 217.589
MDL Number MFCD00021280
InChI Key SIPHWXREAZVVNS-UHFFFAOYSA-N
Synonym cyclohexyltrichlorosilane, trichloro cyclohexyl silane, silane, trichlorocyclohexyl, cyclohexane, trichlorosilyl, cyclohexane, 1-trichlorosilyl, cyclohexyl trichlorosilane, trichlorocyclohexyl silane, silane, cyclohexyltrichloro, 1-trichlorosilyl cyclohexane, trichlorocyclohexylsilane
PubChem CID 60987
IUPAC Name trichloro(cyclohexyl)silane
SMILES C1CCC(CC1)[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 g
UN Number 1763
Formula Weight 217.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyltrichlorosilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.