Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexyltriphenylphosphonium bromide, 98+%

Catalog No. AAA1429822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429822 100 g
AAA1429806 5 g
AAA1429814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429822 Supplier Thermo Scientific Chemicals Supplier No. A1429822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7333-51-9 | C24H26BrP | 425.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7333-51-9
Molecular Formula C24H26BrP
Molecular Weight (g/mol) 425.35
MDL Number MFCD00031667
InChI Key QRAKRDZMIONMRZ-UHFFFAOYSA-M
Synonym cyclohexyltriphenylphosphonium bromide, cyclohexyl triphenylphosphonium bromide, cyclohexyltriphenylphosphanium bromide, cyclohexyl triphenyl phosphanium bromide, acmc-20alcd, cyclohexyltriphenylphosphoniumbromide, cyclohexyl triphenyl phosphonium bromide
PubChem CID 2724861
SMILES [Br-].C1CCC(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 265°C to 272°C
Quantity 100 g
Beilstein 3581012
Sensitivity Hygroscopic
IUPAC Name cyclohexyl(triphenyl)phosphanium;bromide
Formula Weight 425.36
Percent Purity ≥98%
Chemical Name or Material Cyclohexyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-834-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.