Learn More
Selleck Chemical LLC Cycloleucine-E1771-250MG
SDP
Supplier:  Selleck Chemical LLC E1771250MG
Encompass_Preferred
Cycloleucine is a specific antagonist of NMDA receptor(N-methyl-D-aspartate receptor coupled glycine receptor) with a Ki of 600 M Cycloleucine also acts as a specific inhibitor of S-adenosyl-methionine-mediated methylation Cycloleucine exhibits cytostatic effects
Catalog No. 50-313-8928