Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylzirconium(IV) Trichloride 96.0+%, TCI America™
SDP

Catalog No. C19951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19951G 1 g
C19955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19951G Supplier TCI America Supplier No. C19951G

Chemical Identifiers

CAS 34767-44-7
Molecular Formula C5H5Cl3Zr
Synonym Trichloro(cyclopentadienyl)zirconium(IV)

Specifications

Quantity 1 g
UN Number 3261
Formula Weight 262.67
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentadienylzirconium(IV) Trichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.